Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Chloromethyl)Trimethylsilane

Catalog No. 20535425ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-535-425ML 4 x 25 mL
20-535-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 20-535-425ML Supplier Sigma Aldrich Supplier No. 2053544X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(Chloromethyl)trimethylsilane can be treated with aldehydes or ketones in the presence of triphenylphosphine to synthesize terminal alkenes. It can also be used in the preparation of a reagent, trimethylsilylmethyl magnesium chloride, commonly used in Peterson methylenation.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 2344-80-1
Density 0.879 g/mL at 25°C
Boiling Point 98°C to 99°C
Molecular Formula C4H11ClSi
Refractive Index n20/D 1.418
Linear Formula (CH3)3SiCH2Cl
MDL Number MFCD00000878
Quantity 4 x 25 mL
Synonym (Trimethylsilyl)methyl chloride
Molecular Weight (g/mol) 122.67 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.