Learn More
Carbosynth LLC. Chloromethylated Polystyrene Resin (100-200 mesh) 1% DVB, 25g, CAS# [70024-51-0]
SDP
Catalog No. 501686851
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Merrifield Resin, Application: Unsubstituted Resins for Solid Phase Synthesis, Storage Temperature: 2-10C
Catalog No. 50-168-6851 Supplier Carbosynth LLC. Supplier No. RCM1052PI25G
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Merrifield Resin, Application: Unsubstituted Resins for Solid Phase Synthesis, Storage Temperature: 2-10C