Learn More
Chem-Impex International, Inc. Chloromethylated polystyrene (2.0 - 4.5 meq/g, 200 - 400 mesh) | 55844-94-5 | MFCD00146406 | 25G
Catalog No. 50506793
Encompass_Preferred
Chloromethylated polystyrene (2.0 - 4.5 meq/g, 200 - 400 mesh), 55844-94-5, MFCD00146406, 25G
Catalog No. 50-506-793 Supplier Chem-Impex International, Inc. Supplier No. 1247125G
Chloromethylated polystyrene (2.0 - 4.5 meq/g, 200 - 400 mesh), 55844-94-5, MFCD00146406, 25G