Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethylated Polystyrene: 2% Divinylbenzene Copolymer Beads, Thermo Scientific™

Catalog No. AC404750250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404750250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404750250 Supplier Thermo Scientific Chemicals Supplier No. 404750250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70024-51-0
Molecular Formula C12H23NO4
Molecular Weight (g/mol) 245.319
InChI Key FCGHBWUKOGKLKO-UNXYVOJBSA-N
PubChem CID 122172946
IUPAC Name 2-[(3S)-heptan-3-yl]-5-methyl-5-nitro-1,3-dioxane
SMILES CCCCC(CC)C1OCC(CO1)(C)[N+](=O)[O-]

Specifications

Color White
Infrared Spectrum Authentic
Mesh Size 200 to 300 mesh
Quantity 25 g
Physical Form Fine Powder
Chemical Name or Material Chloromethylated polystyrene: 2% divinylbenzene copolymer beads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.