Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Chloromethylene)Dimethyliminium Chloride

Catalog No. 2809095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-090-95G 5 g
18-989-825ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-090-95G Supplier Sigma Aldrich Supplier No. 2809095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(Chloromethylene)Dimethyliminium Chloride

Specifications

CAS 3724-43-4
Melting Point 132°C
Molecular Formula C3H7Cl2N
Linear Formula ClCH=N(CH3)2Cl
MDL Number MFCD00011868
Quantity 5 g
Synonym (Chlormethylene)dimethylammonium chloride; Arnold's reagent; Dimethylchloroformiminium chloride; Vilsmeier reagent
Molecular Weight (g/mol) 128 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.