Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl phenyl sulfide, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
7205-91-6
C7H7ClS
158.65
MFCD00000921
LLSMWLJPWFSMCP-UHFFFAOYSA-N
chloromethyl phenyl sulfide,chloromethylthiobenzene,alpha-chlorothioanisole,phenyl chloromethyl sulfide,chloromethyl phenyl sulphide,phenylthio methyl chloride,chloromethyl thio benzene,sulfide, chloromethyl phenyl,chloromethyl phenyl sulfane,benzene, chloromethyl thio
81623
chloromethylsulfanylbenzene
C1=CC=C(C=C1)SCCl
7205-91-6
C7H7ClS
158.65
MFCD00000921
LLSMWLJPWFSMCP-UHFFFAOYSA-N
chloromethyl phenyl sulfide,chloromethylthiobenzene,alpha-chlorothioanisole,phenyl chloromethyl sulfide,chloromethyl phenyl sulphide,phenylthio methyl chloride,chloromethyl thio benzene,sulfide, chloromethyl phenyl,chloromethyl phenyl sulfane,benzene, chloromethyl thio
81623
chloromethylsulfanylbenzene
C1=CC=C(C=C1)SCCl
Specific Gravity 1.23
Boiling Point 66°C (0.3 hPa)
Quantity 5g
Refractive Index 1.595
Packaging Glass bottle
Color Colorless to Yellow
Density 1.2300g/mL
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula C6H5SCH2Cl
Beilstein 06,III,1002
Fieser 09,107; 06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327
Formula Weight 158.65
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Chloromethyl phenyl sulfide
Show More Show Less

EINECSNumber : 230-579-7