Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chloroneb 100g | 2675-77-6 | 100 G
SDP

Supplier:  Medchemexpress LLC HY116992100G

Encompass_Preferred

Chloroneb is an analytical/reference standard of the fungicide chloroneb (CAS 2675-77-6) supplied for laboratory use in qualitative and quantitative analyses by HPLC, GC, and MS. The 100 g laboratory pack is listed at 95.0% purity.

  • Analytical reference standard for HPLC, GC, and MS.
  • Intended for pesticide residue analysis and method development.
  • Offered at 95.0% purity for the 100 g laboratory pack.
  • Available in multiple lab-scale packaging to match analytical needs.
  • Molecular weight 207.05 g/mol; chemical formula C8H8Cl2O2.

Catalog No. 50-002-28982


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.