Learn More
Strem, An Ascensus Company CAS# 12257-42-0. 1g. Chloronorbornadiene rhodium(I) dimer, 99%. MFCD00198060
SDP
Supplier:  Strem, An Ascensus Company 4505001G
Encompass
CAS# 12257-42-0. 1g. Chloronorbornadiene rhodium(I) dimer, 99%. MFCD00198060. Molecular Weight: 461.00. Molecular Formula: [RhCl(C7H8)]2. Color/form: yellow to orange pwdr. Strem# 45-0500. http://www.strem.com/catalog/v/45-0500/
Catalog No. 50-901-12156