Learn More
Strem, An Ascensus Company CAS# 92361-49-4. Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II), 99%. MFCD09038426. 1G
SDP
Supplier:  Strem, An Ascensus Company 4401171G
Encompass
CAS# 92361-49-4. 1G. Chloro(pentamethylcyclopentadienyl)bis(triphenylphosphine)ruthenium(II), 99%. MFCD09038426. Molecular Formula: C46H45ClP2Ru. Molecular Weight: 796.32. Color/Form: orange pwdr. Stability: store cold (-18°C). Strem#: 44-0117 www.strem.com/catalog/v3/44-0117/
Catalog No. 50-245-461