Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloropentamethyldisilane 96.0+%, TCI America™
SDP

Catalog No. C34315ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34315ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C34315ML Supplier TCI America Supplier No. C34315ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1560-28-7
Molecular Formula C5H15ClSi2
Molecular Weight (g/mol) 166.80
MDL Number MFCD00456692
InChI Key GJCAUTWJWBFMFU-UHFFFAOYSA-N
Synonym 1-Chloro-1,1,2,2,2-pentamethyldisilane
PubChem CID 73795
IUPAC Name 1-chloro-1,1,2,2,2-pentamethyldisilane
SMILES C[Si](C)(C)[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 mL
UN Number 2986
Formula Weight 166.80
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloropentamethyldisilane
EINECSNumber (2)-2040
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.