Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chlorophenol Red sodium salt

Catalog No. AAL1036406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1036406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1036406 Supplier Thermo Scientific Chemicals Supplier No. L1036406
  • $70.60
    $66.65 / Each of 1

    Save $3.95

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 123333-64-2 | C19H11Cl2O5S | 422.25 g/mol

Chlorophenol Red sodium salt is used in the standard samples to indicate pH. A stain used to detect chlorine dioxide in drinking water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorophenol Red sodium salt is used in the standard samples to indicate pH. A stain used to detect chlorine dioxide in drinking water.

Solubility
Soluble in water.

Notes
Transition interval: pH 4.8
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-64-2
Molecular Formula C19H11Cl2O5S
Molecular Weight (g/mol) 422.25
MDL Number MFCD00151199
InChI Key QXTPRQZMDKBTAI-UNOMPAQXSA-M
Synonym chlorphenol red sodium salt
PubChem CID 131846007
IUPAC Name 2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol;sodium
SMILES OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O

Specifications

Melting Point ∽267°C (decomposition)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 445.26
Chemical Name or Material Chlorophenol Red sodium salt
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-619-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.