Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorophosphonazo III, For Spectrophotometric Det. of Alkaline Earth Metals, MilliporeSigma™ Supelco™

Catalog No. 111013414 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3414 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3414 Supplier MilliporeSigma Supelco Supplier No. 260491GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00042698
Synonym Bis(4-chloro-2-phosphonobenzolazo)chromotropic acid

Specifications

CAS 1914-99-4
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C22H16 Cl2N4O14P2S2
Quantity 1 g
Formula Weight 757.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.