Learn More
Hach Company Chlorophosphonazo Indicator Solution for ULR Hardness, 500 mL
Catalog No. 501996283 Shop All Hach Company Products
Encompass
For ultra low range hardness determination by the Chlorophosphonazo method. Hach Method 8374. Range: 4 - 1000 ug/L Ca & Mg as CaCO3. Ideal for use with Hach's Pour-Thru Cell. Requires CDTA Reagent, Cat. No. 2589636. 500 mL bottle
Catalog No. 50-199-6283 Supplier Hach Company Supplier No. 2589549
For ultra low range hardness determination by the Chlorophosphonazo method. Hach Method 8374. Range: 4 - 1000 ug/L Ca & Mg as CaCO3. Ideal for use with Hach's Pour-Thru Cell. Requires CDTA Reagent, Cat. No. 2589636. 500 mL bottle