Learn More
Hach Company Chlorophosphonazo Indicator Solution Pillows for ULR Hardness, pk/100
Catalog No. 501996284 Shop All Hach Company Products
Encompass
For ultra low range hardness determination by the Chlorophosphonazo method. Hach Method 8374. Range: 4 - 1000 ug/L Ca & Mg as CaCO3. Chlorophosphonazo solution reagent packaged in solution pillows. Requires CDTA Reagent, Cat. No. 2589636. 100 Tests.
Catalog No. 50-199-6284 Supplier Hach Company Supplier No. 2589599
For ultra low range hardness determination by the Chlorophosphonazo method. Hach Method 8374. Range: 4 - 1000 ug/L Ca & Mg as CaCO3. Chlorophosphonazo solution reagent packaged in solution pillows. Requires CDTA Reagent, Cat. No. 2589636. 100 Tests.