Learn More
Medchemexpress LLC Chlorophyll a | 479-61-8 | MFCD00079050 | 98.0% | 893.49 g/mol | C55H72MgN4O5 | 1 MG
SDP
Catalog No. 5000282602
Encompass_Preferred
Chlorophyll a is the primary green pigment used in oxygenic photosynthesis research. Supplied as a high-purity biochemical reagent, it is intended for laboratory studies of photosynthetic mechanisms, antioxidant activity, and related biochemical assays. The material is provided in small pack sizes for analytical and experimental applications and includes supporting documentation for handling and quality control.
  • High purity (98%) suitable for research use.
  • Molecular weight 893.49 g/mol and formula C55H72MgN4O5.
  • Provided in a 1 mg pack size for analytical applications.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Store protected from light at -20°C; in solvent store at -80°C up to 6 months.
Catalog No. 50-002-82602 Supplier Medchemexpress LLC Supplier No. HYW1277441MG
Chlorophyll a is the primary green pigment used in oxygenic photosynthesis research. Supplied as a high-purity biochemical reagent, it is intended for laboratory studies of photosynthetic mechanisms, antioxidant activity, and related biochemical assays. The material is provided in small pack sizes for analytical and experimental applications and includes supporting documentation for handling and quality control.
  • High purity (98%) suitable for research use.
  • Molecular weight 893.49 g/mol and formula C55H72MgN4O5.
  • Provided in a 1 mg pack size for analytical applications.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Store protected from light at -20°C; in solvent store at -80°C up to 6 months.