Learn More
Sigma Aldrich Fine Chemicals Biosciences Chlorophyll a from spinach10MG
Catalog No. 502907499
Encompass
Chlorophyll a is an important green or blue color pigment in most organisms which is implicated in oxygenicnbspphotosynthesis. It functions as an antenna pigment in photosystems I and II.
Catalog No. 50-290-7499 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C575310MG
Chlorophyll a is an important green or blue color pigment in most organisms which is implicated in oxygenicnbspphotosynthesis. It functions as an antenna pigment in photosystems I and II.