Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorophyll A, Sigma-Aldrich

Catalog No. 01258861 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-861 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-861 Supplier MilliporeSigma Supelco Supplier No. C61441MG
Add to Cart
Add to Cart

Chlorophyll a is an essential pigment in oxygenic photosynthesis. In photosystems, it is present as an antenna pigment as well as in reaction center I and II. Chlorophyll a is a blue-green pigment which absorbs light in 400 to 450 nm and 640 to 660 nm range. In photosynthesis, the energy absorbed by chlorophyll transforms carbon dioxide and water into carbohydrates and oxygen. When chlorophyll absorbs light energy, an electron in chlorophyll is excited from a lower energy state to a higher energy state. In this higher energy state, the electron is more readily transferred to another molecule. This starts a chain of electron-transfer steps, which ends with an electron transferred to carbon dioxide.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-61-8
Molecular Weight (g/mol) 893.49 g/mol

Specifications

Packaging Vacuum-sealed clear ampule
Quantity 1 mg
Chemical Name or Material Chlorophyll A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.