Learn More
Supelco Inc Chlorophyll a, from Spinach, 1 MG Ampule
Catalog No. NC9759373
Encompass
appearance (Color) Blue to Very Dark Blue appearance (Form) Powder Solubility (Color) Blue, Green, Purity (HPLC) >_ 85 % % Purity (HPLC) <_ 0.4 479-61-8
Catalog No. NC9759373 Supplier Supelco Inc Supplier No. C57531MG
appearance (Color) Blue to Very Dark Blue appearance (Form) Powder Solubility (Color) Blue, Green, Purity (HPLC) >_ 85 % % Purity (HPLC) <_ 0.4 479-61-8