Learn More
Medchemexpress LLC Chlorophyll b | 519-62-0 | MFCD00079053 | 95.0% | 908.48 g·mol⁻¹ | C55H71MgN4O6+ | 500 UG
SDP
Catalog No. 5000283637
Encompass_Preferred
Chlorophyll b is a naturally occurring tetrapyrrole pigment used in plant physiology and biochemical research. It functions in light harvesting for photosynthesis and exhibits antioxidant activity, making it suitable for studies of photosynthetic mechanisms, oxidative stress, and protective effects against DNA damage.
  • Natural tetrapyrrole pigment for photosynthesis research.
  • Antioxidant activity; may increase glutathione levels.
  • Useful in assays for oxidative stress and DNA damage protection.
  • Available in microgram and milligram research sizes for flexible dosing.
  • High purity suitable for laboratory assays and analytical work.
  • Molecular data provided to support experimental calculations.
Catalog No. 50-002-83637 Supplier Medchemexpress LLC Supplier No. HYW127709500UG
Chlorophyll b is a naturally occurring tetrapyrrole pigment used in plant physiology and biochemical research. It functions in light harvesting for photosynthesis and exhibits antioxidant activity, making it suitable for studies of photosynthetic mechanisms, oxidative stress, and protective effects against DNA damage.
  • Natural tetrapyrrole pigment for photosynthesis research.
  • Antioxidant activity; may increase glutathione levels.
  • Useful in assays for oxidative stress and DNA damage protection.
  • Available in microgram and milligram research sizes for flexible dosing.
  • High purity suitable for laboratory assays and analytical work.
  • Molecular data provided to support experimental calculations.