Learn More
Medchemexpress LLC Chlorophyllin sodium copper salt | 11006-34-1 | 95.0% | 1 G
SDP
Catalog No. 5000409345
Encompass_Preferred
Chlorophyllin sodium copper salt is an orally active green pigment that acts as an antioxidant and antimutagenic agent. It also promotes hematopoiesis and thrombopoiesis. This product is for research use only and is not sold to patients.
  • Orally active green pigment
  • Antioxidant
  • Antimutagenic agent
  • Promotes hematopoiesis and thrombopoiesis
Catalog No. 50-004-09345 Supplier Medchemexpress LLC Supplier No. HYB09111G
Chlorophyllin sodium copper salt is an orally active green pigment that acts as an antioxidant and antimutagenic agent. It also promotes hematopoiesis and thrombopoiesis. This product is for research use only and is not sold to patients.
  • Orally active green pigment
  • Antioxidant
  • Antimutagenic agent
  • Promotes hematopoiesis and thrombopoiesis