Learn More
Bioworld Chloroplast Isolation Kit, 20 Prep(s)
Catalog No. 50254004
Encompass_Preferred
Chloroplast isolation kitbioWORLD's Chloroplast isolation kit provides a simple, rapid and efficient procedure to isolate intact chloroplasts from plant leaves. The chloroplast isolation method includes homogenization of the plant leaf tissue followed by isolation of chloroplasts by filtration and finally the separation of intact chloroplast from the broken ones using a percoll gradient centrifugation.
Catalog No. 50-254-004 Supplier Bioworld Supplier No. 302100001
Chloroplast isolation kitbioWORLD's Chloroplast isolation kit provides a simple, rapid and efficient procedure to isolate intact chloroplasts from plant leaves. The chloroplast isolation method includes homogenization of the plant leaf tissue followed by isolation of chloroplasts by filtration and finally the separation of intact chloroplast from the broken ones using a percoll gradient centrifugation.