Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroplatinic Acid, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002894 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-894 1 g Glass Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-894 Supplier Avantor Supplier No. 289003
Add to Cart
Add to Cart

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Packaging Glass Ampoule
Quantity 1 g
Specific Gravity 1L = 2.43kg
Formula Weight 517.92
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystals
Chemical Name or Material Chloroplatinic Acid, 6-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.