Learn More
Strem, An Ascensus Company Chloroplatinicacidhydrate3N 5g
SDP
Catalog No. 5090113200
Encompass
CAS# 18497-13-7. 5g. Chloroplatinic acid hexahydrate (38-40% Pt) (99.9%-Pt). MFCD00149910. Molecular Weight: 409.82 (517.92). Molecular Formula: H2PtCl6∙6H2O. Color/form: orange pwdr. Strem# 78-0200. http://www.strem.com/catalog/v/78-0200/
Catalog No. 50-901-13200 Supplier Strem, An Ascensus Company Supplier No. 7802005G
CAS# 18497-13-7. 5g. Chloroplatinic acid hexahydrate (38-40% Pt) (99.9%-Pt). MFCD00149910. Molecular Weight: 409.82 (517.92). Molecular Formula: H2PtCl6∙6H2O. Color/form: orange pwdr. Strem# 78-0200. http://www.strem.com/catalog/v/78-0200/