Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroplatinic Acid Hexahydrate, ACS Reagent, ≥37.50% Pt basis, Solstice

Catalog No. 60045127 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-127 5 g Glass Bottle
60-045-125 1 g Glass Bottle
60-045-126 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-127 Supplier Solstice Supplier No. 2060835G

missing translation for 'validMainframeMsg'

ACS Reagent,≥37.50% Pt basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Physical Form Powder
Quantity 5 g
Linear Formula H2PtCl6 · 6H2O
UN Number UN2507
Formula Weight 517.9g/mol
Percent Purity ≥37.50% (Pt basis)
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Chloroplatinic Acid Hexahydrate
Hazard Statement P261-P280-P301 + P310-P305 + P351 + P338-P310
Precautionary Statement H301-H314-H317-H334
DOTInformation 8/PGIII
EINECSNumber 241-010-7
RTECSNumber TP1510000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.