Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroplatinic Acid Hydrate 99.9% minimum (Trace Metal Grade), Reagents

Catalog No. 01338045
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-045 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-045 Supplier Reagents Holdings Llc Supplier No. C11760005C2
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloroplatinic Acid Hydrate 99.9% minimum (Trace Metal Grade), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym Hydrogen Hexachloroplatinate (IV), Platinic Chloride
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 115°C
Color Orange
Physical Form Solid
Quantity 5 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 2.431
Chemical Name or Material Chloroplatinic Acid Hydrate 99.9% minimum (Trace Metal Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.