Learn More
Medchemexpress LLC Chloropretadalafil 25g
SDP
Catalog No. 5000205712
Encompass_Preferred
Chloropretadalafil (Compound 31) is a key intermediate that can be used to synthesize Tadalafil (HY-90009A) derivatives[1]
Catalog No. 50-002-05712 Supplier Medchemexpress LLC Supplier No. 25GHYW098292
Chloropretadalafil (Compound 31) is a key intermediate that can be used to synthesize Tadalafil (HY-90009A) derivatives[1]