Learn More
Medchemexpress LLC Chloroprocaine hydrochloride | 3858-89-7 | MFCD00025215 | 99.9% | 10 MM / 1 ML
SDP
Catalog No. 5000413410
Encompass_Preferred
Chloroprocaine hydrochloride (2-Chloroprocaine hydrochloride) is a potent inhibitor of Na,K-ATPase activity with an IC50 of 13 mM. It blocks peripheral nerves. This product is for research use only.
  • Potent inhibitor of Na,K-ATPase activity
  • Blocks peripheral nerve
  • IC50 of 13 mM for Na,K-ATPase
Catalog No. 50-004-13410 Supplier Medchemexpress LLC Supplier No. HYB160410MM/1ML
Chloroprocaine hydrochloride (2-Chloroprocaine hydrochloride) is a potent inhibitor of Na,K-ATPase activity with an IC50 of 13 mM. It blocks peripheral nerves. This product is for research use only.
  • Potent inhibitor of Na,K-ATPase activity
  • Blocks peripheral nerve
  • IC50 of 13 mM for Na,K-ATPase