Learn More
Selleck Chemical LLC Chloropyramine hydrochloride
SDP
Catalog No. 5000003189
Encompass_Preferred
Chloropyramine hydrochloride (Halopyramine) is a histamine receptor H1 antagonist and has anti-cholinergic antispasmodic and antiallergic effects
Catalog No. 50-000-03189 Supplier Selleck Chemical LLC Supplier No. S567025MG
Chloropyramine hydrochloride (Halopyramine) is a histamine receptor H1 antagonist and has anti-cholinergic antispasmodic and antiallergic effects