Learn More
Description
Specifications
Specifications
| Chemical Name or Material | Chloropyramine Hydrochloride |
| CAS | 6170-42-9 |
| Physical Form | Neat |
| Linear Formula | C16H20 ClN3 · HCl |
| Technique | Gas Chromatography |
| MDL Number | MFCD00079009 |
| Synonym | N-p-Chlorobenzyl-N′,N′-dimethyl-N-(2-pyridyl)ethylenediamine |
| Type | Analytical Standard |
| Molecular Weight (g/mol) | 326.26 |
| Industry Type | Drug Analysis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.