Learn More
Medchemexpress LLC Chloropyrazine | 14508-49-7 | MFCD00006124 | 99.9% | 114.53 g/mol | C4H3ClN2 | 25g
SDP
Catalog No. 5000270099
Encompass_Preferred
Chloropyrazine (Pyrazin-2-yl chloride) is a pyrazine-based compound that can be used for the synthesis of active compounds Additionally Chloropyrazine can be used as an electrolyte additive in the research of lithium-sulfur batteries[1][2]
Catalog No. 50-002-70099 Supplier Medchemexpress LLC Supplier No. HYY010925G
Chloropyrazine (Pyrazin-2-yl chloride) is a pyrazine-based compound that can be used for the synthesis of active compounds Additionally Chloropyrazine can be used as an electrolyte additive in the research of lithium-sulfur batteries[1][2]