Learn More
Medchemexpress LLC HY-17589A 100mg Medchemexpress, Chloroquine CAS:54-05-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17589A100MG
Encompass_Preferred
Medchemexpress, HY-17589A 100mg Chloroquine CAS:54-05-7 Chloroquine is an antimalarial and anti-inflammatory agent widely used to treat malaria and rheumatoid arthritis. Chloroquine is an autophagy and toll-like receptors (TLRs) inhibitor. Chloroquine is highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM)[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9245