Learn More
Medchemexpress LLC Chloroquine-d5 | 1854126-41-2 | 99.9% | 324.90 | 1 MG
SDP
Catalog No. 5000387527
Encompass_Preferred
Chloroquine-d5 is a deuterium labeled variant of Chloroquine, an antimalarial and anti-inflammatory agent. It is recognized for its effectiveness against malaria and rheumatoid arthritis, and it also acts as an inhibitor for autophagy and toll-like receptors (TLRs). This compound has shown high efficacy in controlling SARS-CoV-2 (COVID-19) infection in vitro.
  • Deuterium labeled compound
  • Antimalarial and anti-inflammatory properties
  • Inhibitor of autophagy and toll-like receptors (TLRs)
  • Effective in SARS-CoV-2 (COVID-19) infection control in vitro
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-87527 Supplier Medchemexpress LLC Supplier No. HY17589AS1MG
Chloroquine-d5 is a deuterium labeled variant of Chloroquine, an antimalarial and anti-inflammatory agent. It is recognized for its effectiveness against malaria and rheumatoid arthritis, and it also acts as an inhibitor for autophagy and toll-like receptors (TLRs). This compound has shown high efficacy in controlling SARS-CoV-2 (COVID-19) infection in vitro.
  • Deuterium labeled compound
  • Antimalarial and anti-inflammatory properties
  • Inhibitor of autophagy and toll-like receptors (TLRs)
  • Effective in SARS-CoV-2 (COVID-19) infection control in vitro
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS