Learn More
Medchemexpress LLC Chloroquine dihydrochloride | 3545-67-3 | 99.6% | 1 ML
SDP
Catalog No. 5000387507
Encompass_Preferred
Chloroquine dihydrochloride is an antimalarial and anti-inflammatory agent. It is widely used to treat malaria and rheumatoid arthritis, and functions as an autophagy and toll-like receptors (TLRs) inhibitor. This product has shown high effectiveness in controlling SARS-CoV-2 (COVID-19) infection in vitro.
  • Antimalarial and anti-inflammatory agent
  • Used to treat malaria and rheumatoid arthritis
  • Autophagy and toll-like receptors (TLRs) inhibitor
  • Effective in controlling SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM)
  • For research use only
Catalog No. 50-003-87507 Supplier Medchemexpress LLC Supplier No. HY17589B10MM/1ML
Chloroquine dihydrochloride is an antimalarial and anti-inflammatory agent. It is widely used to treat malaria and rheumatoid arthritis, and functions as an autophagy and toll-like receptors (TLRs) inhibitor. This product has shown high effectiveness in controlling SARS-CoV-2 (COVID-19) infection in vitro.
  • Antimalarial and anti-inflammatory agent
  • Used to treat malaria and rheumatoid arthritis
  • Autophagy and toll-like receptors (TLRs) inhibitor
  • Effective in controlling SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM)
  • For research use only