Learn More
Medchemexpress LLC Chloroquine dihydrochloride | 3545-67-3 | 99.61% | 392.79 | 50 MG
SDP
Catalog No. 5000387523
Encompass_Preferred
Chloroquine dihydrochloride is an antimalarial and anti-inflammatory agent widely used to treat malaria and rheumatoid arthritis. It acts as an autophagy and toll-like receptors (TLRs) inhibitor. It is also highly effective in controlling SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM).
  • Antimalarial agent
  • Anti-inflammatory agent
  • Autophagy inhibitor
  • Toll-like receptors (TLRs) inhibitor
  • Effective in controlling SARS-CoV-2 (COVID-19) infection in vitro
Catalog No. 50-003-87523 Supplier Medchemexpress LLC Supplier No. HY17589B50MG
Chloroquine dihydrochloride is an antimalarial and anti-inflammatory agent widely used to treat malaria and rheumatoid arthritis. It acts as an autophagy and toll-like receptors (TLRs) inhibitor. It is also highly effective in controlling SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM).
  • Antimalarial agent
  • Anti-inflammatory agent
  • Autophagy inhibitor
  • Toll-like receptors (TLRs) inhibitor
  • Effective in controlling SARS-CoV-2 (COVID-19) infection in vitro