Learn More
Medchemexpress LLC Chloroquine (dihydrochloride) | 3545-67-3 | MFCD06804666 | 99.6% | 392.79 g/mol | C18H28Cl3N3 | 10 MG
SDP
Catalog No. 5000297960
Encompass_Preferred
Chloroquine (dihydrochloride) is the dihydrochloride salt of chloroquine, a 4-aminoquinoline antimalarial and anti-inflammatory agent used as a research reagent. It is commonly used to inhibit autophagy, alter endosomal/lysosomal pH, and as a screening compound in antiviral and antiparasitic assays.
  • High chemical purity suitable for research (reported ~99.6%).
  • Solid form suitable for formulation or solution preparation.
  • Commonly used as an autophagy inhibitor in cell biology experiments.
  • Modulates endosomal and lysosomal pH for trafficking and uptake studies.
  • Available in small pack sizes for screening and assay workflows.
Catalog No. 50-002-97960 Supplier Medchemexpress LLC Supplier No. HY17589B10MG
Chloroquine (dihydrochloride) is the dihydrochloride salt of chloroquine, a 4-aminoquinoline antimalarial and anti-inflammatory agent used as a research reagent. It is commonly used to inhibit autophagy, alter endosomal/lysosomal pH, and as a screening compound in antiviral and antiparasitic assays.
  • High chemical purity suitable for research (reported ~99.6%).
  • Solid form suitable for formulation or solution preparation.
  • Commonly used as an autophagy inhibitor in cell biology experiments.
  • Modulates endosomal and lysosomal pH for trafficking and uptake studies.
  • Available in small pack sizes for screening and assay workflows.