Learn More
Medchemexpress LLC HY-17589 100mg , Chloroquine (diphosphate) CAS:50-63-5 Purity:98%
SDP
Catalog No. 501492702
Encompass_Preferred
Medchemexpress, HY-17589 100mg Chloroquine (diphosphate) CAS:50-63-5 Formula:C18H32ClN3O8P2 Purity:98% Chloroquine (diphosphate) is an antimalarial drug and autophagy/lysosome inhibitor, inhibiting Toll-like receptor ( TLR ) signalling. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2702 Supplier Medchemexpress LLC Supplier No. HY17589100MG
Medchemexpress, HY-17589 100mg Chloroquine (diphosphate) CAS:50-63-5 Formula:C18H32ClN3O8P2 Purity:98% Chloroquine (diphosphate) is an antimalarial drug and autophagy/lysosome inhibitor, inhibiting Toll-like receptor ( TLR ) signalling. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.