Learn More
Selleck Chemical LLC Chloroquine diphosphate S4157-200mg
SDP
Catalog No. 503108381
Encompass_Preferred
Chloroquine diphosphate is a 4-aminoquinoline anti-malarial and anti-rheumatoid agent also acting as an ATM activator Chloroquine is also an inhibitor of toll-like receptors (TLRs)
Catalog No. 50-310-8381 Supplier Selleck Chemical LLC Supplier No. S4157200MG
Chloroquine diphosphate is a 4-aminoquinoline anti-malarial and anti-rheumatoid agent also acting as an ATM activator Chloroquine is also an inhibitor of toll-like receptors (TLRs)