Learn More
AdipoGen Chloroquine diphosphate
SDP
Supplier:  AdipoGen AGCR13721G025
Encompass_Preferred
Chemical. CAS 50-63-5. Formula C18H26ClN3 . 2H3PO4. MW 319.9 . 196.0. Chloroquine is a commonly used antimalarial drug, more toxic than its derivative hydroxychloroquine Prod. No. AG-CR1-3720. Chloroquine has anti-inflammatory, immunomodulating, anti-infective, antiviral, antithrombotic and metabolic effects. It has anticancer properties, related to their strong antiproliferative, antimutagenic, epigenetic and autophagy inhibiting and apoptosis inducing activities. Chloroquine is used to treat rheumatoid arthritis, systemic lupus erythematosus, antiphospholipid antibody syndrome and Sjögrens syndrome. Chloroquine interfers with lysosomal activity and autophagy, interacts with membrane stability and alters signaling pathways and transcriptional activity, which can result in inhibition of cytokine production and modulation of certain co-stimulatory molecules.
Catalog No. 50-256-2194