Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroquine diphosphate sa250G
Catalog No. 502903446
Encompass
Chloroquine is a member of quinolone family and is a weak intercalating agent. Chloroquine is used for treating amebiasis rheumatoid arthritis discoid and systemic lupus erythematosus.
Catalog No. 50-290-3446 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C6628250G
Chloroquine is a member of quinolone family and is a weak intercalating agent. Chloroquine is used for treating amebiasis rheumatoid arthritis discoid and systemic lupus erythematosus.