Learn More
Medchemexpress LLC Chloroquine phosphate | 50-63-5 | 99.9% | 500 MG
SDP
Catalog No. 5000387119
Encompass_Preferred
Chloroquine phosphate is an antimalarial and anti-inflammatory agent commonly used to treat malaria and rheumatoid arthritis. It acts as an inhibitor of autophagy and toll-like receptors (TLRs). Additionally, it has shown high effectiveness in controlling SARS-CoV-2 (COVID-19) infection in vitro, with an EC50 of 1.13 μM. This product is intended for research use only and is not sold to patients.
  • Antimalarial agent
  • Anti-inflammatory agent
  • Autophagy inhibitor
  • TLR inhibitor
  • SARS-CoV-2 efficacy (EC50=1.13 μM)
Catalog No. 50-003-87119 Supplier Medchemexpress LLC Supplier No. HY17589500MG
Chloroquine phosphate is an antimalarial and anti-inflammatory agent commonly used to treat malaria and rheumatoid arthritis. It acts as an inhibitor of autophagy and toll-like receptors (TLRs). Additionally, it has shown high effectiveness in controlling SARS-CoV-2 (COVID-19) infection in vitro, with an EC50 of 1.13 μM. This product is intended for research use only and is not sold to patients.
  • Antimalarial agent
  • Anti-inflammatory agent
  • Autophagy inhibitor
  • TLR inhibitor
  • SARS-CoV-2 efficacy (EC50=1.13 μM)