Learn More
Medchemexpress LLC Chloroquine phosphate | 50-63-5 | MFCD00069852 | 99.7% | 515.86 g/mol | C18H32ClN3O8P2 | 10 MG
SDP
Catalog No. 5000298762
Encompass_Preferred
Chloroquine phosphate (standard) is an analytical reference material supplied as a white to off-white solid for research and quality-control applications. It has a reported purity of 99.67% and a molecular weight of 515.86 g/mol, and is intended for use in assay calibration, method development, and stability testing.
  • High purity suitable for analytical use.
  • Solid form, white to off-white appearance.
  • Intended for assay calibration and method development.
  • Available in small pack sizes for laboratory testing.
  • Includes CAS identifier 50-63-5 for unambiguous identification.
Catalog No. 50-002-98762 Supplier Medchemexpress LLC Supplier No. HY17589R10MG
Chloroquine phosphate (standard) is an analytical reference material supplied as a white to off-white solid for research and quality-control applications. It has a reported purity of 99.67% and a molecular weight of 515.86 g/mol, and is intended for use in assay calibration, method development, and stability testing.
  • High purity suitable for analytical use.
  • Solid form, white to off-white appearance.
  • Intended for assay calibration and method development.
  • Available in small pack sizes for laboratory testing.
  • Includes CAS identifier 50-63-5 for unambiguous identification.