Learn More
Selleck Chemical LLC Chloroquine diphosphate 50mg 50-63-5
SDP
Supplier:  Selleck Chemical LLC S415750MG
Encompass_Preferred
Chloroquine diphosphate is a 4-aminoquinoline anti-malarial and anti-rheumatoid agent, also acting as an ATM activator. Chloroquine is also an inhibitor of toll-like receptors (TLRs). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0941720