Learn More
TARGETMOL CHEMICALS INC Chloroquinoxaline sulfonamide
SDP
Catalog No. 502876406
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Chloroquinoxaline sulfonamide (Chloroquinoxaline) is a topoisomerase II alpha/beta toxin with antitumor activity and immunosuppressive properties that inhibits the proliferation of B16 melanoma cells in mice, and can be used to study metastatic colorectal cancer. It can be used to study metastatic colorectal cancer. Purity 98.67%
Catalog No. 50-287-6406 Supplier TARGETMOL CHEMICALS INC Supplier No. T149535MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Chloroquinoxaline sulfonamide (Chloroquinoxaline) is a topoisomerase II alpha/beta toxin with antitumor activity and immunosuppressive properties that inhibits the proliferation of B16 melanoma cells in mice, and can be used to study metastatic colorectal cancer. It can be used to study metastatic colorectal cancer. Purity 98.67%