Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorosulfonic Acid

Catalog No. 5710245G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 x 100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-102-45G -
57-102-4100G -
57-102-41KG -
57-102-4 4 x 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 57-102-45G Supplier Sigma Aldrich Supplier No. 5710245G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorosulfonic acid is a strong acid that is synthesized on an industrial scale by reacting sulfur trioxide and dry hydrogen chloride gas in an equimolar ratio. It can act as a sulfonating, dehydrating, oxidizing and chlorinating agent. It is reported to be a better electrophilic olefin cyclization agent in comparison to other sulfonic acids. Its infrared spectra in the solid, liquid and gaseous state have been recorded in the region 4000-400cm-1.Chlorosulfonic acid may be used in the following processes: Synthesis of di-n-butyl ammonium chlorosulfonate, a secondary amine ionic liquid.To prepare sulfonated polyethersulfone (SPES) with enhanced hydrophilic property.To attach sulfate group to silk fibroin for improving its anticoagulant ability.As a catalyst to prepare substituted coumarin, via von Pechmann condensation between phenol and +a-ketoester.As a substitute to acid catalyst in Biginelli reaction to prepare 3,4dihydropyrimidines.

Specifications

CAS 7790-94-5
Molecular Formula HClO3S
Linear Formula ClSO3H
MDL Number MFCD00011523
Molecular Weight (g/mol) 116.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.