Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorosulfonyl isocyanate

Catalog No. 6913991KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-139-91KG 1 kg
69-139-910KG 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-139-91KG Supplier Sigma Aldrich Supplier No. 6913991KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorosulfonyl isocyanate

Specifications

CAS 1189-71-5
Melting Point -44°C (lit.)
Density 1.626 g/mL (at 25°C (literature))
Boiling Point 107°C (lit.)
Molecular Formula CClNO3S
Refractive Index n20/D 1.447 (literature)
Linear Formula ClSO2NCO
MDL Number MFCD00011608
Quantity 1 kg
Synonym N-Carbonylsulfamyl chloride; CSI
Molecular Weight (g/mol) 141.53
Percent Purity 99.0-100.3% (w/w) (T)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.