Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorosulfonyl isocyanate, 98+%, Thermo Scientific™

Catalog No. AC162040250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC162040250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162040250 Supplier Thermo Scientific Chemicals Supplier No. 162040250

missing translation for 'validMainframeMsg'

Alkylation reagent, C-X bond formation reagent

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189-71-5
Molecular Formula CClNO3S
Molecular Weight (g/mol) 141.525
InChI Key WRJWRGBVPUUDLA-UHFFFAOYSA-N
Synonym chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
PubChem CID 70918
IUPAC Name N-(oxomethylidene)sulfamoyl chloride
SMILES C(=NS(=O)(=O)Cl)=O

Specifications

Melting Point -44.0°C
Density 1.6260g/mL
Boiling Point 106.0°C
Flash Point >110°C
Refractive Index 1.4450 to 1.4490
Linear Formula ClSO2NCO
Quantity 25 mL
Solubility Information Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons, miscible with saturated and chlorinated, hydrocarbons
Formula Weight 141.53
Chemical Name or Material Chlorosulfonyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.