Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorosulfonylacetyl chloride

Catalog No. 2597721G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-977-21G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-977-21G Supplier Sigma Aldrich Supplier No. 2597721G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorosulfonylacetyl chloride

Specifications

CAS 4025-77-8
Density 1.669 g/mL (at 25°C (literature))
Boiling Point 71°C to 72°C (1 mmHg)
Molecular Formula C2H2Cl2O3S
Refractive Index n20/D 1.493 (literature)
Linear Formula ClSO2CH2COCl
MDL Number MFCD00007457
Quantity 100 mg
Molecular Weight (g/mol) 177.01
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.