Learn More
Cayman Chemical Chlorothalonil, Purity: ≥98%, CAS: 1897-45-6, 100 mg
SDP
Catalog No. 502935511
Encompass_Preferred
A broad-spectrum organochlorine fungicide; forms adducts with GST and enzyme cysteine residues leading to GST depletion and enzyme deactivation; inhibits the growth of fungi as well as Gram-positive and Gram-negative bacteria (MICs = 0.5-1.7 µg/ml); completely inhibits the growth of P. infestans, the tomato late blight pathogen, on tomato plants at 100 µg/ml; toxic to aquatic organisms, including species of fish, crustaceans, molluscs, and algae with tenth percentile of toxicity values of 25.23, 40.59, 0.69, and 3.94 µg/L, respectively; carcinogenic in animal models and induces neoplasms in the forestomach and kidneys of rats at 3.8 mg/kg per day, but it is not genotoxic
Catalog No. 50-293-5511 Supplier Cayman Chemical Supplier No. 24142100 MG
A broad-spectrum organochlorine fungicide; forms adducts with GST and enzyme cysteine residues leading to GST depletion and enzyme deactivation; inhibits the growth of fungi as well as Gram-positive and Gram-negative bacteria (MICs = 0.5-1.7 µg/ml); completely inhibits the growth of P. infestans, the tomato late blight pathogen, on tomato plants at 100 µg/ml; toxic to aquatic organisms, including species of fish, crustaceans, molluscs, and algae with tenth percentile of toxicity values of 25.23, 40.59, 0.69, and 3.94 µg/L, respectively; carcinogenic in animal models and induces neoplasms in the forestomach and kidneys of rats at 3.8 mg/kg per day, but it is not genotoxic