Learn More
Medchemexpress LLC Chlorothalonil | 1897-45-6 | 250 MG
SDP
Catalog No. 5000421563
Encompass_Preferred
Chlorothalonil is a broad-spectrum foliar fungicide with oral activity, effective against fungal diseases in vegetable and crop leaves. It can influence the microbial community in soil, inhibit spermatogenesis, and potentially cause intestinal epithelial barrier dysfunction and fetal toxicity. This product is intended for research use only.
  • Broad-spectrum foliar fungicide
  • Exhibits oral activity
  • Combats fungal diseases in vegetable and crop leaves
  • Can alter the microbial community in the soil
  • May inhibit spermatogenesis
  • Potential to cause intestinal epithelial barrier dysfunction and fetal toxicity
Catalog No. 50-004-21563 Supplier Medchemexpress LLC Supplier No. 250MGHYN6625
Chlorothalonil is a broad-spectrum foliar fungicide with oral activity, effective against fungal diseases in vegetable and crop leaves. It can influence the microbial community in soil, inhibit spermatogenesis, and potentially cause intestinal epithelial barrier dysfunction and fetal toxicity. This product is intended for research use only.
  • Broad-spectrum foliar fungicide
  • Exhibits oral activity
  • Combats fungal diseases in vegetable and crop leaves
  • Can alter the microbial community in the soil
  • May inhibit spermatogenesis
  • Potential to cause intestinal epithelial barrier dysfunction and fetal toxicity