Learn More
Medchemexpress LLC Chlorothalonil | 1897-45-6 | MFCD00045594 | 250 MG
SDP
Catalog No. 5000422081
Encompass_Preferred
Chlorothalonil is a broad-spectrum foliar fungicide with oral activity. It can be used to combat fungal diseases in vegetable and crop leaves. It can alter the microbial community in the soil. Chlorothalonil inhibits spermatogenesis, and can cause intestinal epithelial barrier dysfunction and fetal toxicity.
  • Broad-spectrum foliar fungicide
  • Oral activity
  • Can be used to combat fungal diseases in vegetable and crop leaves
Catalog No. 50-004-22081 Supplier Medchemexpress LLC Supplier No. 250MGHYN6625R
Chlorothalonil is a broad-spectrum foliar fungicide with oral activity. It can be used to combat fungal diseases in vegetable and crop leaves. It can alter the microbial community in the soil. Chlorothalonil inhibits spermatogenesis, and can cause intestinal epithelial barrier dysfunction and fetal toxicity.
  • Broad-spectrum foliar fungicide
  • Oral activity
  • Can be used to combat fungal diseases in vegetable and crop leaves